Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGTCTGGCCTGGAAGTAAACGAC[A/G]TCCCTGATGCGCTCTTTGCAGTCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606948 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANAPC5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANAPC5 - anaphase promoting complex subunit 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001137559.1 | 2258 | Silent Mutation | GAC,GAT | D,D 600 | NP_001131031.1 | |
NM_016237.4 | 2258 | Silent Mutation | GAC,GAT | D,D 712 | NP_057321.2 | |
XM_005253900.3 | 2258 | Silent Mutation | GAC,GAT | D,D 699 | XP_005253957.1 | |
XM_017019423.1 | 2258 | Silent Mutation | GAC,GAT | D,D 314 | XP_016874912.1 | |
XM_017019424.1 | 2258 | Silent Mutation | GAC,GAT | D,D 314 | XP_016874913.1 |