Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCTCTGTTTTAGAAATGAAGTCA[C/T]GACTAGGAATTGGGAGCGAGGGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606949 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANAPC7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANAPC7 - anaphase promoting complex subunit 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001137664.1 | 1727 | Intron | NP_001131136.1 | |||
NM_016238.2 | 1727 | UTR 3 | NP_057322.2 | |||
XM_005253903.3 | 1727 | UTR 3 | XP_005253960.1 | |||
XM_011538454.2 | 1727 | UTR 3 | XP_011536756.1 | |||
XM_017019425.1 | 1727 | UTR 3 | XP_016874914.1 | |||
XM_017019426.1 | 1727 | UTR 3 | XP_016874915.1 | |||
XM_017019427.1 | 1727 | UTR 3 | XP_016874916.1 | |||
XM_017019428.1 | 1727 | UTR 3 | XP_016874917.1 |