Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCTGACGCGGGCGCTGGCGCTGGC[C/G]CTGGTGCTGGCCCTGCTGGTCGGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
PLBD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PLBD2 - phospholipase B domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159727.1 | 107 | Silent Mutation | GCC,GCG | A,A 24 | NP_001153199.1 | |
NM_173542.3 | 107 | Silent Mutation | GCC,GCG | A,A 24 | NP_775813.2 | |
XM_011538023.2 | 107 | Intron | XP_011536325.1 | |||
XM_017018977.1 | 107 | UTR 5 | XP_016874466.1 |