Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGGGCACCCAGCACAGTGTCTC[A/G]CACACAGCAGATGCCCAGCAGGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616444 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBM19 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RBM19 - RNA binding motif protein 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146698.1 | 3821 | Intron | NP_001140170.1 | |||
NM_001146699.1 | 3821 | UTR 3 | NP_001140171.1 | |||
NM_016196.3 | 3821 | Intron | NP_057280.2 | |||
XM_017020280.1 | 3821 | UTR 3 | XP_016875769.1 | |||
XM_017020281.1 | 3821 | Intron | XP_016875770.1 |