Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTCCCACAGCAAACCAAGAAAAA[C/T]AGACACACTGGCTGCCTGCCTAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602654 MIM: 604984 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FARP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FARP1 - FERM, ARH/RhoGEF and pleckstrin domain protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STK24 - serine/threonine kinase 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032296.3 | 2988 | UTR 3 | NP_001027467.2 | |||
NM_001286649.1 | 2988 | UTR 3 | NP_001273578.1 | |||
NM_003576.4 | 2988 | UTR 3 | NP_003567.2 | |||
XM_005254079.4 | 2988 | Intron | XP_005254136.1 | |||
XM_017020794.1 | 2988 | Intron | XP_016876283.1 | |||
XM_017020795.1 | 2988 | Intron | XP_016876284.1 |