Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTTTCTTACCAAAACTGATTTAT[A/G]CGCCTTAATTCTATGTACAATATTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TEX30 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TEX30 - testis expressed 30 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286775.1 | 486 | Missense Mutation | CAT,TAT | H,Y 78 | NP_001273704.1 | |
NM_001286776.1 | 486 | Missense Mutation | CAT,TAT | H,Y 37 | NP_001273705.1 | |
NM_138779.4 | 486 | Missense Mutation | CAT,TAT | H,Y 78 | NP_620134.3 | |
XM_005254097.2 | 486 | Missense Mutation | CAT,TAT | H,Y 78 | XP_005254154.1 | |
XM_011521135.2 | 486 | Missense Mutation | CAT,TAT | H,Y 78 | XP_011519437.1 | |
XM_011521136.2 | 486 | Intron | XP_011519438.1 | |||
XM_017020852.1 | 486 | Missense Mutation | CAT,TAT | H,Y 78 | XP_016876341.1 |