Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGACTCTGTGAGCAGTTTCCACTC[G/T]GCGCCCAAGCGCTTGCTGATCTCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604974 | ||||||||||||||||||||
Literature Links: |
LOC101927248 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC101927248 - uncharacterized LOC101927248 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SOX21 - SRY-box 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007084.3 | 626 | Silent Mutation | GCA,GCC | A,A 44 | NP_009015.1 |
SOX21-AS1 - SOX21 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |