Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTTTTGAGGTATTTCATAATACA[A/T]GTTGGGTATTTTAAATAGTAAAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608671 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLDN10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLDN10 - claudin 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DZIP1 - DAZ interacting zinc finger protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014934.4 | 4370 | UTR 3 | NP_055749.1 | |||
NM_198968.3 | 4370 | UTR 3 | NP_945319.1 | |||
XM_005254032.1 | 4370 | Intron | XP_005254089.1 | |||
XM_006719916.1 | 4370 | Intron | XP_006719979.1 | |||
XM_006719917.2 | 4370 | UTR 3 | XP_006719980.1 | |||
XM_006719918.1 | 4370 | Intron | XP_006719981.1 | |||
XM_006719920.1 | 4370 | Intron | XP_006719983.1 | |||
XM_017020444.1 | 4370 | Intron | XP_016875933.1 | |||
XM_017020445.1 | 4370 | Intron | XP_016875934.1 | |||
XM_017020446.1 | 4370 | Intron | XP_016875935.1 | |||
XM_017020447.1 | 4370 | Intron | XP_016875936.1 |