Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCTCAAATCAATCTAGAAGGAGA[A/C]ATTAGGAATAACCAGGAAAATGTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605431 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAPK8IP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAPK8IP3 - mitogen-activated protein kinase 8 interacting protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040439.1 | Intron | NP_001035529.1 | ||||
NM_001318852.1 | Intron | NP_001305781.1 | ||||
NM_015133.4 | Intron | NP_055948.2 | ||||
XM_005255190.2 | Intron | XP_005255247.1 | ||||
XM_006720869.2 | Intron | XP_006720932.1 | ||||
XM_011522429.2 | Intron | XP_011520731.1 | ||||
XM_011522430.2 | Intron | XP_011520732.1 | ||||
XM_011522431.2 | Intron | XP_011520733.1 | ||||
XM_011522432.2 | Intron | XP_011520734.1 | ||||
XM_011522433.2 | Intron | XP_011520735.1 | ||||
XM_017023079.1 | Intron | XP_016878568.1 | ||||
XM_017023080.1 | Intron | XP_016878569.1 | ||||
XM_017023081.1 | Intron | XP_016878570.1 |