Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAATATAAGGACTGAAATAAAAGT[A/G]AATTTGAAAGATGGCTAATCTACTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607669 MIM: 607032 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARL6IP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARL6IP1 - ADP ribosylation factor like GTPase 6 interacting protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMG1 - SMG1, nonsense mediated mRNA decay associated PI3K related kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015092.4 | 10021 | UTR 3 | NP_055907.3 | |||
XM_005255182.3 | 10021 | UTR 3 | XP_005255239.1 | |||
XM_005255183.3 | 10021 | UTR 3 | XP_005255240.1 | |||
XM_005255184.3 | 10021 | UTR 3 | XP_005255241.1 | |||
XM_011545769.2 | 10021 | UTR 3 | XP_011544071.1 | |||
XM_011545770.1 | 10021 | UTR 3 | XP_011544072.1 | |||
XM_017023066.1 | 10021 | UTR 3 | XP_016878555.1 | |||
XM_017023067.1 | 10021 | UTR 3 | XP_016878556.1 |