Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGAGACTTGAGTTATAAATTACA[C/T]GATCAGAGGAAAGGACTTAAAATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMC5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMC5 - transmembrane channel like 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105248.1 | Intron | NP_001098718.1 | ||||
NM_001105249.1 | Intron | NP_001098719.1 | ||||
NM_001261841.1 | Intron | NP_001248770.1 | ||||
NM_001308161.1 | Intron | NP_001295090.1 | ||||
NM_024780.4 | Intron | NP_079056.2 |