Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAGCTGCCGTTGGGAAATCCTGC[C/T]GTTCGGAGAGCTGGGGGAGGGAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605948 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPRC5B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPRC5B - G protein-coupled receptor class C group 5 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304771.1 | 1227 | Silent Mutation | ACA,ACG | T,T 478 | NP_001291700.1 | |
NM_016235.2 | 1227 | Silent Mutation | ACA,ACG | T,T 347 | NP_057319.1 | |
XM_006721051.2 | 1227 | Silent Mutation | ACA,ACG | T,T 347 | XP_006721114.1 | |
XM_006721052.2 | 1227 | Silent Mutation | ACA,ACG | T,T 347 | XP_006721115.1 |
IQCK - IQ motif containing K | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |