Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTGCAAATTAGAAACACCCAGAA[A/G]CTTTTAAAATACATGGTAATTCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
USP31 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
USP31 - ubiquitin specific peptidase 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020718.3 | 8224 | UTR 3 | NP_065769.3 | |||
XM_017023496.1 | 8224 | UTR 3 | XP_016878985.1 | |||
XM_017023497.1 | 8224 | UTR 3 | XP_016878986.1 | |||
XM_017023498.1 | 8224 | UTR 3 | XP_016878987.1 | |||
XM_017023499.1 | 8224 | UTR 3 | XP_016878988.1 | |||
XM_017023500.1 | 8224 | Intron | XP_016878989.1 |