Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCTGTCGGTCATTGAGAATGGGA[C/T]GGACACTGGGCTGCTGCAGCCGGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603750 | ||||||||||||||||||||
Literature Links: |
AQP8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AQP8 - aquaporin 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001169.2 | 264 | Missense Mutation | ACG,ATG | T,M 61 | NP_001160.2 | |
XM_011545822.2 | 264 | Missense Mutation | ACG,ATG | T,M 62 | XP_011544124.1 | |
XM_011545823.2 | 264 | Missense Mutation | ACG,ATG | T,M 62 | XP_011544125.1 |