Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGGGATCGGGCTGCCGACGCCCCG[G/T]GGCAGCGGCACCAACGGCTACGTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606032 | ||||||||||||||||||||
Literature Links: |
SRRM2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SRRM2 - serine/arginine repetitive matrix 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016333.3 | 560 | Silent Mutation | CGG,CGT | R,R 11 | NP_057417.3 | |
XM_005255226.1 | 560 | Silent Mutation | CGG,CGT | R,R 11 | XP_005255283.1 | |
XM_006720874.3 | 560 | Silent Mutation | CGG,CGT | R,R 11 | XP_006720937.1 |
SRRM2-AS1 - SRRM2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |