Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCTGGGTGAATGAGGGCAGGTCC[C/T]GGAAGGCAGGGGCATAGCTGCAGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608411 | ||||||||||||||||||||
Literature Links: |
XPO6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
XPO6 - exportin 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270940.1 | Intron | NP_001257869.1 | ||||
NM_015171.3 | Intron | NP_055986.1 | ||||
XM_005255197.3 | Intron | XP_005255254.1 | ||||
XM_005255198.1 | Intron | XP_005255255.1 | ||||
XM_005255199.2 | Intron | XP_005255256.1 | ||||
XM_011545772.2 | Intron | XP_011544074.1 |