Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGTGCCTCTCTCTTCCTTTCGCA[A/G]CTGCTGCCGCCCACCCCTCGTCTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608937 MIM: 602389 | ||||||||||||||||||||
Literature Links: |
SH2B1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SH2B1 - SH2B adaptor protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145795.1 | 608 | Intron | NP_001139267.1 | |||
NM_001145796.1 | 608 | Intron | NP_001139268.1 | |||
NM_001145797.1 | 608 | Intron | NP_001139269.1 | |||
NM_001145812.1 | 608 | Intron | NP_001139284.1 | |||
NM_001308293.1 | 608 | Intron | NP_001295222.1 | |||
NM_001308294.1 | 608 | Intron | NP_001295223.1 | |||
NM_015503.2 | 608 | UTR 5 | NP_056318.2 | |||
XM_017023114.1 | 608 | UTR 5 | XP_016878603.1 | |||
XM_017023115.1 | 608 | Intron | XP_016878604.1 | |||
XM_017023116.1 | 608 | Intron | XP_016878605.1 | |||
XM_017023117.1 | 608 | UTR 5 | XP_016878606.1 | |||
XM_017023118.1 | 608 | Intron | XP_016878607.1 | |||
XM_017023119.1 | 608 | Intron | XP_016878608.1 | |||
XM_017023120.1 | 608 | Intron | XP_016878609.1 |
TUFM - Tu translation elongation factor, mitochondrial | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |