Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCATGTGCTGGACCAGAACAGCAA[C/T]GTGTCCCGTGTGGAGGTCGGGCCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605088 MIM: 612033 | ||||||||||||||||||||
Literature Links: |
MVP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MVP - major vault protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001293204.1 | 265 | Silent Mutation | AAC,AAT | N,N 24 | NP_001280133.1 | |
NM_001293205.1 | 265 | Silent Mutation | AAC,AAT | N,N 24 | NP_001280134.1 | |
NM_005115.4 | 265 | Silent Mutation | AAC,AAT | N,N 24 | NP_005106.2 | |
NM_017458.3 | 265 | Silent Mutation | AAC,AAT | N,N 24 | NP_059447.2 |
PAGR1 - PAXIP1 associated glutamate rich protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |