Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGTCCGCTTCTGTCGGACACTTC[A/G]CCACCTCCGGATCCTGGGCAGCCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF747 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF747 - zinc finger protein 747 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305018.1 | 748 | Missense Mutation | GCG,GTG | A,V 106 | NP_001291947.1 | |
NM_001305019.1 | 748 | Missense Mutation | GCG,GTG | A,V 106 | NP_001291948.1 | |
NM_001305020.1 | 748 | Nonsense Mutation | CGA,TGA | R,* 153 | NP_001291949.1 | |
NM_023931.3 | 748 | Nonsense Mutation | CGA,TGA | R,* 153 | NP_076420.1 |
ZNF768 - zinc finger protein 768 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |