Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGGCTGAGAGATGAGTCCTGGAT[A/T]TCTGAGTCACTAGAGGGAGCGTTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615549 MIM: 182381 MIM: 602353 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARMC5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARMC5 - armadillo repeat containing 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC5A2 - solute carrier family 5 member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TGFB1I1 - transforming growth factor beta 1 induced transcript 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042454.2 | Intron | NP_001035919.1 | ||||
NM_001164719.1 | Intron | NP_001158191.1 | ||||
NM_015927.4 | Intron | NP_057011.2 |