Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGGGGTCGGGATGGCCCCATCAG[C/T]TCCGCTCCACCTTCTTCTTGCCCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615648 MIM: 613278 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NLRC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NLRC3 - NLR family CARD domain containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLX4 - SLX4 structure-specific endonuclease subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032444.2 | 5533 | Missense Mutation | AAC,AGC | N,S 1834 | NP_115820.2 | |
XM_011522715.2 | 5533 | Missense Mutation | AAC,AGC | N,S 1833 | XP_011521017.1 | |
XM_017023775.1 | 5533 | Missense Mutation | AAC,AGC | N,S 1560 | XP_016879264.1 |