Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGAATTCACCCAAGTTCTAGCC[A/G]CAGAGGCCAGGCTGCCCAGACGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608738 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAB11FIP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAB11FIP3 - RAB11 family interacting protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142272.1 | Intron | NP_001135744.1 | ||||
NM_014700.3 | Intron | NP_055515.1 | ||||
XM_005255713.3 | Intron | XP_005255770.1 | ||||
XM_005255714.1 | Intron | XP_005255771.1 | ||||
XM_005255715.4 | Intron | XP_005255772.1 | ||||
XM_005255717.3 | Intron | XP_005255774.1 | ||||
XM_005255718.3 | Intron | XP_005255775.1 | ||||
XM_011522764.2 | Intron | XP_011521066.1 | ||||
XM_011522765.2 | Intron | XP_011521067.1 | ||||
XM_017023907.1 | Intron | XP_016879396.1 |