Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAGGAGTGCAGGGGTGACATGTG[C/T]CGGGGCCAGAGAGGTATCTTCCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611668 MIM: 614336 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CORO7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CORO7 - coronin 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201472.1 | 3063 | UTR 3 | NP_001188401.1 | |||
NM_001201473.1 | 3063 | UTR 3 | NP_001188402.1 | |||
NM_024535.4 | 3063 | UTR 3 | NP_078811.3 |
CORO7-PAM16 - CORO7-PAM16 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201479.1 | 3063 | Intron | NP_001188408.1 |
PAM16 - presequence translocase-associated motor 16 homolog (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |