Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGATGGACATATCCTTGTTGACA[C/T]AGCGGTCATACTGAGCAGGGGTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602871 MIM: 609771 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPL - periplakin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002705.4 | 5315 | Missense Mutation | TAT,TGT | Y,C 1738 | NP_002696.3 | |
XM_006720902.3 | 5315 | Missense Mutation | TAT,TGT | Y,C 1751 | XP_006720965.1 | |
XM_017023374.1 | 5315 | Missense Mutation | TAT,TGT | Y,C 1767 | XP_016878863.1 | |
XM_017023375.1 | 5315 | Missense Mutation | TAT,TGT | Y,C 1754 | XP_016878864.1 |
UBN1 - ubinuclein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |