Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCACGTGACATCCACGGAGGGCG[G/T]CCGGGCTTATCTGGTGCTGCGTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613201 MIM: 607298 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHTF18 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHTF18 - chromosome transmission fidelity factor 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022092.2 | 619 | Missense Mutation | GGC,GTC | G,V 186 | NP_071375.1 | |
XM_005255470.1 | 619 | Missense Mutation | GGC,GTC | G,V 70 | XP_005255527.1 | |
XM_005255471.3 | 619 | Missense Mutation | GGC,GTC | G,V 214 | XP_005255528.1 | |
XM_011522572.1 | 619 | Missense Mutation | GGC,GTC | G,V 187 | XP_011520874.1 | |
XM_011522573.1 | 619 | Intron | XP_011520875.1 | |||
XM_017023532.1 | 619 | Missense Mutation | GGC,GTC | G,V 165 | XP_016879021.1 | |
XM_017023533.1 | 619 | Missense Mutation | GGC,GTC | G,V 187 | XP_016879022.1 | |
XM_017023534.1 | 619 | Missense Mutation | GGC,GTC | G,V 70 | XP_016879023.1 |
GNG13 - G protein subunit gamma 13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPUSD1 - RNA pseudouridylate synthase domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |