Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAGGGGTCCTTGGGGATCCCGTC[C/T]TCGATCCACTTCAGCAGCCTGCGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613201 MIM: 607298 | ||||||||||||||||||||
Literature Links: |
CHTF18 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHTF18 - chromosome transmission fidelity factor 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022092.2 | 219 | Intron | NP_071375.1 | |||
XM_005255470.1 | 219 | Intron | XP_005255527.1 | |||
XM_005255471.3 | 219 | Intron | XP_005255528.1 | |||
XM_011522572.1 | 219 | Intron | XP_011520874.1 | |||
XM_011522573.1 | 219 | Intron | XP_011520875.1 | |||
XM_017023532.1 | 219 | Intron | XP_016879021.1 | |||
XM_017023533.1 | 219 | Intron | XP_016879022.1 | |||
XM_017023534.1 | 219 | Intron | XP_016879023.1 |
GNG13 - G protein subunit gamma 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016541.2 | 219 | Silent Mutation | NP_057625.1 |
PRR25 - proline rich 25 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |