Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCCACAACAGAGACGCCTGGGGA[C/T]GCCACTTTGTGCAGCGGGCGCTTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613395 MIM: 600968 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR138-2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR138-2 - microRNA 138-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC12A3 - solute carrier family 12 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000339.2 | 65 | Silent Mutation | GAC,GAT | D,D 12 | NP_000330.2 | |
NM_001126107.1 | 65 | Silent Mutation | GAC,GAT | D,D 12 | NP_001119579.1 | |
NM_001126108.1 | 65 | Silent Mutation | GAC,GAT | D,D 12 | NP_001119580.1 | |
XM_005256119.2 | 65 | Silent Mutation | GAC,GAT | D,D 12 | XP_005256176.1 |