Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTGGGAAGATGCAGAGGCTCATG[A/G]TGCTCCTCGCCACATCGGGCGCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601120 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDH5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDH5 - cadherin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001795.4 | 164 | Missense Mutation | ATG,GTG | M,V 6 | NP_001786.2 | |
XM_011522801.1 | 164 | Missense Mutation | ATG,GTG | M,V 6 | XP_011521103.1 |