Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAGTTCCCGCTAAACCGGCAGGC[A/G]ATGAGAAAGGAAACCATCACTAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CES4A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CES4A - carboxylesterase 4A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190201.1 | 1303 | Silent Mutation | GCA,GCG | A,A 272 | NP_001177130.1 | |
NM_001190202.1 | 1303 | Silent Mutation | GCA,GCG | A,A 276 | NP_001177131.1 | |
NM_001318506.1 | 1303 | Silent Mutation | GCA,GCG | A,A 176 | NP_001305435.1 | |
NM_173815.6 | 1303 | Silent Mutation | GCA,GCG | A,A 370 | NP_776176.5 | |
XM_005255896.4 | 1303 | Silent Mutation | GCA,GCG | A,A 393 | XP_005255953.1 | |
XM_011523021.2 | 1303 | Silent Mutation | GCA,GCG | A,A 437 | XP_011521323.1 | |
XM_011523023.2 | 1303 | Silent Mutation | GCA,GCG | A,A 433 | XP_011521325.1 | |
XM_011523024.2 | 1303 | Silent Mutation | GCA,GCG | A,A 437 | XP_011521326.1 | |
XM_011523025.2 | 1303 | Intron | XP_011521327.1 | |||
XM_011523026.2 | 1303 | Silent Mutation | GCA,GCG | A,A 397 | XP_011521328.1 | |
XM_011523027.2 | 1303 | Silent Mutation | GCA,GCG | A,A 437 | XP_011521329.1 | |
XM_011523028.2 | 1303 | Silent Mutation | GCA,GCG | A,A 385 | XP_011521330.1 | |
XM_011523029.2 | 1303 | Silent Mutation | GCA,GCG | A,A 437 | XP_011521331.1 | |
XM_011523030.2 | 1303 | Intron | XP_011521332.1 | |||
XM_017023160.1 | 1303 | Intron | XP_016878649.1 | |||
XM_017023161.1 | 1303 | Silent Mutation | GCA,GCG | A,A 433 | XP_016878650.1 | |
XM_017023162.1 | 1303 | Intron | XP_016878651.1 | |||
XM_017023163.1 | 1303 | Intron | XP_016878652.1 | |||
XM_017023164.1 | 1303 | Intron | XP_016878653.1 | |||
XM_017023165.1 | 1303 | Silent Mutation | GCA,GCG | A,A 433 | XP_016878654.1 | |
XM_017023166.1 | 1303 | Intron | XP_016878655.1 | |||
XM_017023167.1 | 1303 | Silent Mutation | GCA,GCG | A,A 272 | XP_016878656.1 | |
XM_017023168.1 | 1303 | Intron | XP_016878657.1 |