Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCTATAGCCCTCATCATTTCAAG[A/C]CAAATTTTAAAATCTCAAAATGTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613202 MIM: 602428 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHTF8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHTF8 - chromosome transmission fidelity factor 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039690.3 | Intron | NP_001034779.1 | ||||
NM_001040146.3 | Intron | NP_001035236.1 | ||||
XM_011523167.1 | Intron | XP_011521469.1 | ||||
XM_011523168.1 | Intron | XP_011521470.1 | ||||
XM_017023370.1 | Intron | XP_016878859.1 |
HAS3 - hyaluronan synthase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |