Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTGGTGGGGAGAGCCCATTTACC[A/G]GAATGGGCAGTATGTTGGCAAGACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC400541 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC400541 - uncharacterized LOC400541 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDPR - pyruvate dehydrogenase phosphatase regulatory subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322117.1 | 2979 | Missense Mutation | CAG,CGG | Q,R 791 | NP_001309046.1 | |
NM_001322118.1 | 2979 | Missense Mutation | CAG,CGG | Q,R 691 | NP_001309047.1 | |
NM_001322119.1 | 2979 | Missense Mutation | CAG,CGG | Q,R 389 | NP_001309048.1 | |
NM_017990.4 | 2979 | Missense Mutation | CAG,CGG | Q,R 791 | NP_060460.4 | |
XM_005256015.2 | 2979 | Missense Mutation | CAG,CGG | Q,R 791 | XP_005256072.1 | |
XM_005256016.3 | 2979 | Missense Mutation | CAG,CGG | Q,R 791 | XP_005256073.1 | |
XM_006721224.2 | 2979 | Missense Mutation | CAG,CGG | Q,R 730 | XP_006721287.1 | |
XM_006721225.3 | 2979 | Intron | XP_006721288.1 | |||
XM_011523190.2 | 2979 | Intron | XP_011521492.1 | |||
XM_017023387.1 | 2979 | Missense Mutation | CAG,CGG | Q,R 791 | XP_016878876.1 | |
XM_017023388.1 | 2979 | Intron | XP_016878877.1 | |||
XM_017023389.1 | 2979 | Intron | XP_016878878.1 |