Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGAAAGCTCAGGTCTGGAGAGCCG[G/T]TATCTATAGATGCGAGCGTGACGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601065 MIM: 606490 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AARS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AARS - alanyl-tRNA synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001605.2 | 3268 | UTR 3 | NP_001596.2 |
EXOSC6 - exosome component 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_058219.2 | 3268 | Intron | NP_478126.1 |
LOC107984138 - serine/threonine-protein kinase SMG1-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |