Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGAGGACAGAGCTGCCCAGTCCT[C/T]ACTCAACAAGCTGATCAGAAGCAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
DDX19A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DDX19A - DEAD-box helicase 19A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320522.1 | 342 | Missense Mutation | TCA,TTA | S,L 60 | NP_001307451.1 | |
NM_001320525.1 | 342 | UTR 5 | NP_001307454.1 | |||
NM_001320526.1 | 342 | UTR 5 | NP_001307455.1 | |||
NM_001320527.1 | 342 | Intron | NP_001307456.1 | |||
NM_018332.4 | 342 | Missense Mutation | TCA,TTA | S,L 60 | NP_060802.1 |
LOC100506083 - uncharacterized LOC100506083 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |