Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCGGTTATTCTCTCCAGTCACTCA[C/T]CTGTTACCTTAATCATTCTTTTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612060 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNRF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNRF1 - zinc and ring finger 1, E3 ubiquitin protein ligase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032268.4 | 1755 | Intron | NP_115644.1 | |||
XM_011523392.1 | 1755 | Intron | XP_011521694.1 | |||
XM_017023793.1 | 1755 | Intron | XP_016879282.1 | |||
XM_017023794.1 | 1755 | UTR 3 | XP_016879283.1 |