Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTTAACAGAGTTTTTGGTTGTTA[C/T]CCTAAAGTTTAGTGGAACTTTAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZFP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZFP1 - ZFP1 zinc finger protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318469.1 | Intron | NP_001305398.1 | ||||
NM_001318471.1 | Intron | NP_001305400.1 | ||||
NM_001318472.1 | Intron | NP_001305401.1 | ||||
NM_001318473.1 | Intron | NP_001305402.1 | ||||
NM_001318474.1 | Intron | NP_001305403.1 | ||||
NM_001318475.1 | Intron | NP_001305404.1 | ||||
NM_001318476.1 | Intron | NP_001305405.1 | ||||
NM_153688.3 | Intron | NP_710155.2 | ||||
XM_011522921.2 | Intron | XP_011521223.1 | ||||
XM_017022983.1 | Intron | XP_016878472.1 | ||||
XM_017022984.1 | Intron | XP_016878473.1 | ||||
XM_017022985.1 | Intron | XP_016878474.1 | ||||
XM_017022986.1 | Intron | XP_016878475.1 | ||||
XM_017022987.1 | Intron | XP_016878476.1 |