Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCCACTGTTGTTTCTAGAACTTT[A/G]TCAACATAGTCCACACGATCAGGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601501 | ||||||||||||||||||||
Literature Links: |
VPS35 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
VPS35 - VPS35 retromer complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018206.5 | 1209 | Silent Mutation | GAC,GAT | D,D 370 | NP_060676.2 | |
XM_005256045.3 | 1209 | Silent Mutation | GAC,GAT | D,D 303 | XP_005256102.1 | |
XM_011523227.2 | 1209 | Silent Mutation | GAC,GAT | D,D 341 | XP_011521529.1 |