Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGGCAGGCAAATTATTCAGCCAC[G/T]CGTGTTTCAGGCACTGTGTGGCACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612147 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYLK3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYLK3 - myosin light chain kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308301.1 | 1600 | Missense Mutation | GAG,GCG | E,A 427 | NP_001295230.1 | |
NM_182493.2 | 1600 | Missense Mutation | GAG,GCG | E,A 768 | NP_872299.2 | |
XM_006721334.3 | 1600 | Missense Mutation | GAG,GCG | E,A 427 | XP_006721397.1 |