Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATTGTGCGCTGTGTGATGACTCA[A/G]TTCTTCTGCTAACCTTTCTCTCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613082 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP2C2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP2C2 - ATPase secretory pathway Ca2+ transporting 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286527.2 | Intron | NP_001273456.2 | ||||
NM_001291454.1 | Intron | NP_001278383.1 | ||||
NM_014861.3 | Intron | NP_055676.3 | ||||
XM_011523486.2 | Intron | XP_011521788.1 | ||||
XM_011523487.2 | Intron | XP_011521789.1 |