Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATGTTCGGAGGGGGTCTGGTGCT[C/T]AAACGTTCATTCTGGGCTTCCTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600385 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADCY7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADCY7 - adenylate cyclase 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
BRD7 - bromodomain containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001173984.2 | 1938 | Intron | NP_001167455.1 | |||
NM_013263.4 | 1938 | Intron | NP_037395.2 | |||
XM_011523046.2 | 1938 | Intron | XP_011521348.1 | |||
XM_011523047.2 | 1938 | Silent Mutation | TTA,TTG | L,L 581 | XP_011521349.1 | |
XM_011523048.2 | 1938 | Intron | XP_011521350.1 | |||
XM_011523049.2 | 1938 | Intron | XP_011521351.1 | |||
XM_011523050.2 | 1938 | Silent Mutation | TTA,TTG | L,L 363 | XP_011521352.1 | |
XM_017023179.1 | 1938 | Intron | XP_016878668.1 | |||
XM_017023180.1 | 1938 | Intron | XP_016878669.1 | |||
XM_017023181.1 | 1938 | Silent Mutation | TTA,TTG | L,L 362 | XP_016878670.1 |