Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAGGAGTTTGTCTTCTCTTTCAA[C/T]GCCATGGCGTCCTCTTCCATGCACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614975 MIM: 601089 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FENDRR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FENDRR - FOXF1 adjacent non-coding developmental regulatory RNA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FOXF1 - forkhead box F1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001451.2 | 1090 | Silent Mutation | AAC,AAT | N,N 349 | NP_001442.2 |