Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCAGAGGCAAATGTGACCCGCCT[C/T]CTACGCAGGAGGCAACAGAGGCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616820 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FLJ30679 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FLJ30679 - uncharacterized protein FLJ30679 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FOXC2-AS1 - FOXC2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MTHFSD - methenyltetrahydrofolate synthetase domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159377.1 | Intron | NP_001152849.1 | ||||
NM_001159378.1 | Intron | NP_001152850.1 | ||||
NM_001159379.1 | Intron | NP_001152851.1 | ||||
NM_001159380.1 | Intron | NP_001152852.1 | ||||
NM_022764.2 | Intron | NP_073601.2 |