Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGATGCCATGCCTGCTCCCCCAG[C/T]CTAATGGGCTTTGATGGGGGAAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605956 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NOD2 - nucleotide binding oligomerization domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001293557.1 | 92 | Intron | NP_001280486.1 | |||
NM_022162.2 | 92 | UTR 5 | NP_071445.1 | |||
XM_005256084.3 | 92 | Intron | XP_005256141.1 | |||
XM_006721242.3 | 92 | Intron | XP_006721305.1 | |||
XM_006721243.3 | 92 | Intron | XP_006721306.1 | |||
XM_011523259.2 | 92 | Intron | XP_011521561.1 | |||
XM_011523260.2 | 92 | Intron | XP_011521562.1 | |||
XM_011523261.1 | 92 | Intron | XP_011521563.1 | |||
XM_017023535.1 | 92 | Intron | XP_016879024.1 | |||
XM_017023536.1 | 92 | Intron | XP_016879025.1 | |||
XM_017023537.1 | 92 | Intron | XP_016879026.1 | |||
XM_017023538.1 | 92 | Intron | XP_016879027.1 |