Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCACACATCTCTGGACGTGGTGG[A/G]TGAGAAGATCTCCGCAATGGGGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612222 MIM: 610970 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GALNS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GALNS - galactosamine (N-acetyl)-6-sulfatase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PABPN1L - poly(A) binding protein nuclear 1 like (cytoplasmic) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRAPPC2L - trafficking protein particle complex 2-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318524.1 | 184 | Missense Mutation | GAT,GGT | D,G 40 | NP_001305453.1 | |
NM_001318525.1 | 184 | Missense Mutation | GAT,GGT | D,G 40 | NP_001305454.1 | |
NM_001318526.1 | 184 | Missense Mutation | ATG,GTG | M,V 1 | NP_001305455.1 | |
NM_001318527.1 | 184 | UTR 5 | NP_001305456.1 | |||
NM_001318528.1 | 184 | Missense Mutation | GAT,GGT | D,G 10 | NP_001305457.1 | |
NM_001318529.1 | 184 | UTR 5 | NP_001305458.1 | |||
NM_001318530.1 | 184 | Missense Mutation | GAT,GGT | D,G 10 | NP_001305459.1 | |
NM_001318532.1 | 184 | UTR 5 | NP_001305461.1 | |||
NM_016209.4 | 184 | Missense Mutation | GAT,GGT | D,G 40 | NP_057293.1 | |
XM_017023270.1 | 184 | Missense Mutation | ATG,GTG | M,V 1 | XP_016878759.1 |