Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATGTCCAGGCGGAAACAGAGCAA[C/T]CCCCGGCAGATCAAGCGTGAGTCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601950 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZFPM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZFPM1 - zinc finger protein, FOG family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153813.2 | 57 | Silent Mutation | AAC,AAT | N,N 8 | NP_722520.2 | |
XM_011522912.2 | 57 | Intron | XP_011521214.1 | |||
XM_011522914.2 | 57 | Intron | XP_011521216.1 | |||
XM_011522917.2 | 57 | Intron | XP_011521219.1 | |||
XM_017022982.1 | 57 | Intron | XP_016878471.1 |