Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCCGGCCGTGCAGGGTGTCCAGG[A/G]GGCCGGCTGCCTGGCCCAGGAACCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114019 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDH15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDH15 - cadherin 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004933.2 | 2770 | Intron | NP_004924.1 |
SLC22A31 - solute carrier family 22 member 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242757.1 | 2770 | Missense Mutation | CCC,CTC | P,L 256 | NP_001229686.1 | |
XM_011522897.2 | 2770 | Missense Mutation | CCC,CTC | P,L 256 | XP_011521199.1 | |
XM_011522899.2 | 2770 | Missense Mutation | CCC,CTC | P,L 256 | XP_011521201.1 | |
XM_017022962.1 | 2770 | Missense Mutation | CCC,CTC | P,L 289 | XP_016878451.1 | |
XM_017022963.1 | 2770 | Missense Mutation | CCC,CTC | P,L 263 | XP_016878452.1 | |
XM_017022964.1 | 2770 | Missense Mutation | CCC,CTC | P,L 263 | XP_016878453.1 | |
XM_017022965.1 | 2770 | Missense Mutation | CCC,CTC | P,L 263 | XP_016878454.1 | |
XM_017022966.1 | 2770 | Missense Mutation | CCC,CTC | P,L 263 | XP_016878455.1 |
ZNF778 - zinc finger protein 778 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |