Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGGAGGTGCTGAGGGAGGAGGGG[C/T]CCAGGGCTCTCCCCAGGCAGCTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105376790 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105376790 - uncharacterized LOC105376790 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OGFOD3 - 2-oxoglutarate and iron dependent oxygenase domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024648.2 | 2603 | UTR 3 | NP_078924.1 | |||
NM_175902.4 | 2603 | UTR 3 | NP_787098.3 |