Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCTCAACTACCGAAAGGGAACAG[A/G]TGCCAGGTGAGACAGCAAGTGGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606676 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRPV2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRPV2 - transient receptor potential cation channel subfamily V member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016113.4 | 593 | Missense Mutation | GAT,GGT | D,G 65 | NP_057197.2 | |
XM_005256676.1 | 593 | Missense Mutation | GAT,GGT | D,G 65 | XP_005256733.1 | |
XM_005256677.1 | 593 | Missense Mutation | GAT,GGT | D,G 65 | XP_005256734.1 | |
XM_005256678.4 | 593 | Missense Mutation | GAT,GGT | D,G 65 | XP_005256735.1 | |
XM_006721541.3 | 593 | Missense Mutation | GAT,GGT | D,G 65 | XP_006721604.1 | |
XM_006721543.3 | 593 | Missense Mutation | GAT,GGT | D,G 65 | XP_006721606.1 | |
XM_011523922.1 | 593 | Missense Mutation | GAT,GGT | D,G 65 | XP_011522224.1 | |
XM_011523923.1 | 593 | Missense Mutation | GAT,GGT | D,G 65 | XP_011522225.1 | |
XM_011523925.2 | 593 | Intron | XP_011522227.1 | |||
XM_017024730.1 | 593 | Missense Mutation | GAT,GGT | D,G 65 | XP_016880219.1 | |
XM_017024731.1 | 593 | Missense Mutation | GAT,GGT | D,G 65 | XP_016880220.1 | |
XM_017024732.1 | 593 | Missense Mutation | GAT,GGT | D,G 65 | XP_016880221.1 |