Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACGTACTCGTCGTCCTCCGTGAT[A/G]AGAACATTCTCCCTGTTGTTCTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607273 MIM: 614960 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FLCN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FLCN - folliculin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLD6 - phospholipase D family member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178836.3 | 607 | Silent Mutation | CTC,CTT | L,L 187 | NP_849158.2 | |
XM_017024310.1 | 607 | Missense Mutation | TCA,TTA | S,L 186 | XP_016879799.1 |