Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTTGCTTTTCTCAACTTCTCAGG[A/G]GAAGGCCATCAGCATTTGGGAGTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605292 | ||||||||||||||||||||
Literature Links: |
NT5M PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NT5M - 5',3'-nucleotidase, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020201.3 | 454 | Missense Mutation | GAG,GGG | E,G 90 | NP_064586.1 | |
XM_005256731.3 | 454 | Missense Mutation | GAG,GGG | E,G 90 | XP_005256788.1 | |
XM_005256732.3 | 454 | Missense Mutation | GAG,GGG | E,G 90 | XP_005256789.1 | |
XM_005256733.3 | 454 | Missense Mutation | GAG,GGG | E,G 90 | XP_005256790.1 | |
XM_005256734.3 | 454 | Missense Mutation | GAG,GGG | E,G 90 | XP_005256791.1 | |
XM_005256737.1 | 454 | UTR 5 | XP_005256794.1 | |||
XM_011523962.2 | 454 | Missense Mutation | GAG,GGG | E,G 90 | XP_011522264.1 | |
XM_011523963.1 | 454 | UTR 5 | XP_011522265.1 | |||
XM_011523964.2 | 454 | UTR 5 | XP_011522266.1 | |||
XM_011523965.2 | 454 | UTR 5 | XP_011522267.1 | |||
XM_011523966.1 | 454 | Missense Mutation | GAG,GGG | E,G 90 | XP_011522268.1 | |
XM_011523967.2 | 454 | Missense Mutation | GAG,GGG | E,G 90 | XP_011522269.1 | |
XM_017024879.1 | 454 | UTR 5 | XP_016880368.1 |