Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGTCCCAAGGTGGCAAGCCTTCAT[A/G]AAGGCACTGCCCAGCTCACAATCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612116 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105369205 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105369205 - uncharacterized LOC105369205 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011524128.2 | 4122 | Intron | XP_011522430.1 | |||
XM_011524129.2 | 4122 | Intron | XP_011522431.1 | |||
XM_011524131.2 | 4122 | UTR 3 | XP_011522433.1 | |||
XM_017025507.1 | 4122 | Intron | XP_016880996.1 | |||
XM_017025508.1 | 4122 | Intron | XP_016880997.1 | |||
XM_017025509.1 | 4122 | Intron | XP_016880998.1 |
USP22 - ubiquitin specific peptidase 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015276.1 | 4122 | UTR 3 | NP_056091.1 | |||
XM_005256575.2 | 4122 | UTR 3 | XP_005256632.1 |